missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescent Compatible Sample Buffer (4X, non-reducing)

Product Code. 17198445
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
Product Code. Quantity unitSize
17198445 5 mL Each
1 options
This item is not returnable. View return policy

Product Code. 17198445

Brand: Invitrogen™ LC2570

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Invitrogen Fluorescent Compatible Sample Buffer is for use in SDS-polyacrylamide gel electrophoresis (SDS-PAGE).

It is optimized for use in fluorescent western blotting as it does not interfere in fluorescent channels, unlike other standard sample buffers. Fluorescent Compatible Sample Buffer is non-reducing but may also be used under denaturing conditions.

Non-reducing—ready-to-use for non-reducing SDS-PAGE, or the preferred type and amount of reducing agent (e.g., DTT) can be added to produce reducing conditions
Lane marker dyes—visible markers of the dye front
Convenient—stable at room temperature, enabling storage at the bench where electrophoresis is performed
Concentrated—4X formulation provides more versatility than traditional 2X buffers
Ideal for fluorescent detection—formulated to prevent auto fluorescence during fluorescent detection

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Buffer Sample Loading Buffers
Concentration 4X
Detection Method Fluorescence
Gel Type Tricine, Tris-Glycine, SDS-PAGE, Bis-Tris, Bis-Tris Plus, Tris-Acetate
Product Type Fluorescent Compatible Sample Buffer
Gel Compatibility SDS-PAGE Gels
Quantity 5 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.