Learn More
Cytiva Fluorinated Propylene Monomer O-Ring
Product Code. 12471090 Shop All Cytiva Products
Change view
Click to view available options
Size:
5.3 x 2.4mm
Unit Size:
Each
Product Code. Size unitSize
12471090 5.3 x 2.4mm Each
1 options
Product Code. 12471090 Supplier Cytiva Supplier No. 18111860
In Stock
Add to basket
Add to basket
Approved to biological reactivity testing in vivo, USP class VI.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Dummy pH electrode
Quantity 1Each
Product Type O-Ring
Material Fully Fluorinated Propylene Monomer
Size 5.3 x 2.4mm