missing translation for 'onlineSavingsMsg'
Learn More

Fluorinert FC 40 (Technical Grade), TRC

Product Code. 30400779
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400779 25 g
30394798 10 g
30393578 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400779 Supplier Toronto Research Chemicals Supplier No. TRCF48580825G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51142-49-5
Molecular Formula C9F21N •C12F27N
Molecular Weight (g/mol) 1192.16
Synonym Electronic Liquid FC 40, FC 40, FC 40 oil, Fluorinert Electronic Liquid FC 40
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N-(perfluorobutyl)-N-(trifluoromethyl)butan-1-amine--tris(perfluorobutyl)amine (1/1)
SMILES FC(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12F27N.C9F21N/c13-1(14,7(25,26)27)4(19,20)10(34,35)40(11(36,37)5(21,22)2(15,16)8(28,29)30)12(38,39)6(23,24)3(17,18)9(31,32)33;10-1(11,5(18,19)20)3(14,15)7(24,25)31(9(28,29)30)8(26,27)4(16,17)2(12,13)6(21,22)23
Formula Weight 1191.93
Chemical Name or Material Fluorinert FC 40 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.