missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Fluoro Plastics/Fluoro Rubber/Glass Superloops

Product Code. 10359227 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10359227 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10359227 Supplier Cytiva Supplier No. 19758501
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Superloop allows the introduction of large sample volumes into a pressurised fluid LC system

Superloop allows the introduction of large sample volumes into a pressurised fluid LC system. It is used together with the injection valve MV-7, INV-907, V9-Inj, or V9H-Inj and replaces a simple sample loop. Superloop works in conjunction with 7-port valves. A movable seal creates two chambers, such that buffer pumped into the upper chamber forces sample from the lower chamber without mixing or diluting the sample. Once the seal reaches the bottom of the lower chamber, a bypass channel in the Superloop 10mL and 50mL or a titanium valve in Superloop 150mL allows the buffer to follow the sample to the column, if desired.
  • Superloop can be used in conjunction with 7-port valves.
  • The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection.
  • They may also be used to inject pulses of a specific buffer solution.
  • Flexible: inject large or small sample volumes.
  • High reproducibility and recovery.
  • No gradient dilution with internal movable seal.
TRUSTED_SUSTAINABILITY

Specifications

Size (Metric) 30 mm dia. x 130 mm L
Max. Pressure 40 Bar
Temperature (Metric) Max 4°C to 40°C
For Use With (Equipment) Used together with preparative chromatography systems from GE Healthcare
Length (Metric) 130 mm
Diameter (Metric) Inner 30 mm
Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.