Learn More
Cytiva AC 10 Flow Adapter
Product Code. 10704762 Shop All Cytiva Products
Change view
Click to view available options
For Use With:
Liquid Chromatography Columns
Unit Size:
Each
Product Code. For Use With unitSize
10704762 Liquid Chromatography Columns Each
1 options
Product Code. 10704762
Brand: Cytiva 19500601
Only null left
Add to basket
Add to basket
Help keep bed height of column constant and protect from insoluble particles
  • Resistant to aqueous solutions and solvents used in liquid chromatography
  • Made from fluoro rubber
TRUSTED_SUSTAINABILITY
Specifications
Type Resistant to Aqueous Solutions and Solvents Used in Liquid Chromatography
For Use With (Application) Liquid Chromatography
Description AC 10 Flow Adapter
For Use With Liquid Chromatography Columns
Material Fluoro Rubber