missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorobenzene, TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | Fluorobenzene |
| CAS | 462-06-6 |
| Molecular Formula | C6 H5 F |
| Quantity | 250 g |
| Synonym | Benzene, fluoro-, Fluorobenzene, Monofluorobenzene, NSC 68416, Phenyl fluoride |
| InChI Formula | InChI=1S/C6H5F/c7-6-4-2-1-3-5-6/h1-5H |
| SMILES | Fc1ccccc1 |
| IUPAC Name | fluorobenzene |
| Molecular Weight (g/mol) | 96.1 |
| Formula Weight | 96.0375 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction