missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoroboric acid, 50 wt% solution in water
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Unit Size:
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Color | White |
| Melting Point | -90.0°C |
| Boiling Point | 130.0°C |
| Density | 1.4100g/mL |
| Packaging | Plastic bottle |
| Specific Gravity | 1.41 |
| Quantity | 25 g |
| Linear Formula | HBF4 |
| Physical Form | Adhering Crystals and/or Chunks |
| UN Number | 1775 |
| Show More |
Safety and Handling
Product Identifier
- Fluoroboric acid, 50 wt% solution in water
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion