missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Fluoxetine hydrochloride

Product Code. 18732724
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732724 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732724 Supplier R&D Systems™ Supplier No. 0927/10
  • ONLINE EXCLUSIVE PRICE
    168.00 €
    158.55 € / 10mg
    Save 9.45 €
    5% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-HT reuptake inhibitor

Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. Binds to the human 5-HT transporter with a Ki of 0.9 nmol/l and is between 150- and 900-fold selective over 5-HT1A, 5-HT2A, H1, α1, α2-adrenergic, and muscarinic receptors. Antidepressant. Induces differentiation of neuronal precursors, enhancing neuronal characteristics. Fluoxetine also inhibits assembly and activation of the NLRP3-ASC inflammasome and prevents degeneration of retinal pigmented epithelium (RPE) cells in an animal model of dry age-related macular degeneration (AMD).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56296-78-7
Molecular Formula C17H19ClF3NO

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 345.78
Purity >99%
Molecular Weight (g/mol) 345.79
Chemical Name or Material Fluoxetine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.