missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine Octanoate, TRC

Product Code. 30389661
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389661 25 mg
30401261 250 mg
30408479 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389661 Supplier Toronto Research Chemicals Supplier No. TRCF59842525MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97671-70-0
Molecular Formula C30H40F3N3O2S
Molecular Weight (g/mol) 563.72
Synonym 2-(4-(3-(2-(Trifluoromethyl)-10H-phenothiazin-10-yl)propyl)piperazin-1-yl)ethyl Octanoate, Fluphenazine EP Impurity D
SMILES CCCCCCCC(=O)OCCN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C30H40F3N3O2S/c1-2-3-4-5-6-12-29(37)38-22-21-35-19-17-34(18-20-35)15-9-16-36-25-10-7-8-11-27(25)39-28-14-13-24(23-26(28)36)30(31,32)33/h7-8,10-11,13-14,23H,2-6,9,12,15-22H2,1H3
Formula Weight 563.2793
Chemical Name or Material Fluphenazine Octanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.