missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine Sulphone, TRC

Product Code. 18091040
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18091040 25 mg 25mg
18001550 100 mg 100mg
18050750 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18091040 Supplier Toronto Research Chemicals Supplier No. TRCF59835025MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1476-79-5
Molecular Formula C22 H26 F3 N3 O3 S
Molecular Weight (g/mol) 469.52
Synonym 1-Piperazineethanol, 4-[3-[5,5-dioxido-2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]- (9CI), 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-, S,S-dioxide, 2-[4-[3-[5,5-Dioxo-2-(trifluoromethyl)-10H-5λ6-phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol, Fluphenazine S,S-Dioxide, Fluphenazine Sulfone
IUPAC Name 2-[4-[3-[5,5-dioxo-2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCCN2c3ccccc3S(=O)(=O)c4ccc(cc24)C(F)(F)F)CC1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H26F3N3O3S/c23-22(24,25)17-6-7-21-19(16-17)28(18-4-1-2-5-20(18)32(21,30)31)9-3-8-26-10-12-27(13-11-26)14-15-29/h1-2,4-7,16,29H,3,8-15H2
Formula Weight 469.1647
Chemical Name or Material Fluphenazine Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.