missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies (±)-Flurbiprofen

Product Code. 17160292
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17160292 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17160292 Supplier STEMCELL Technologies Supplier No. 73924
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PI3K/AKT pathway inhibitor; Inhibits COX-1 and COX-2

  • (±)-Flurbiprofen is a cell-permeable and non-steroidal anti-inflammatory agent
  • It is a potent non-selective inhibitor of cyclooxygenase (COX) with EC50 = 0.1 and 0.4 μM for the inhibition of recombinant human COX-1 and COX-2, respectively (Gierse et al.)
  • Maintains self-renewal and pluripotency in hESC cells grown in conditions that induce differentiation (Bhanu et al.; Desbordes et al.)
  • Inhibits tumor growth in models of prostate cancer, familial adenomatous polyposis, and brain cancer (King, Jr & Khalili; Grubbs et al.; Wechter et al.; Zemskova et al.)

Specifications

CAS 5104-49-4
Synonym Ansaid
Quantity 5 g
Molecular Formula C15H13FO2
Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.