missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flurbiprofen, 98%

Product Code. 15216058 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15216058 1 g 1g
15226058 5 g 5g
15236058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15216058 Supplier Thermo Scientific Chemicals Supplier No. 454660010
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

Flurbiprofen, CAS # 5104-49-4, is a monocarboxylic acid-based nonsteroidal anti-inflammatory (NSAID).

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Flurbiprofen is composed of a 2-fluoro-[1,1'-biphenyl-4-yl] moiety attached to propionic acid

• Flurbiprofen works by inhibiting prostaglandin-endoperoxide synthase and reducing the production of prostaglandins and pyrogens

Applications

• Flurbiprofen is useful for studying pathways associated with prostaglandin production

• It is suitable for laboratory research on pain, inflammation, ocular pressure, and postoperative recovery

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5104-49-4
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00079303
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
Synonym flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
PubChem CID 3394
ChEBI CHEBI:5130
IUPAC Name 2-(3-fluoro-4-phenylphenyl)propanoic acid
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Specifications

Melting Point 110.0°C to 114.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 15,4229
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, readily soluble in most polar solvents
Formula Weight 244.27
Percent Purity 98%
Chemical Name or Material Flurbiprofen
compliance-icons
Product Identifier
  • Flurbiprofen
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.