missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen, TRC

Product Code. 30137082
Change view
Click to view available options
Quantity:
500 mg
1 g
25 g
Unit Size:
1g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137082 25 g 25g
30006552 500 mg 500mg
30011552 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137082 Supplier Toronto Research Chemicals Supplier No. TRCF59870025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5104-49-4
Molecular Formula C15 H13 F O2
Molecular Weight (g/mol) 244.26
Synonym Flurbiprofen, (2RS)-2-(2-Fluorobiphenyl-4-yl)propanoic acid, 4-Biphenylacetic acid, 2-fluoro-α-methyl- (8CI), Hydratropic acid, 3-fluoro-4-phenyl- (7CI), 2-Fluoro-α-methyl[1,1'-biphenyl]-4-acetic acid, (±)-Flurbiprofen, 2-(2-Fluoro-1,1'-biphenyl-4-yl)propanoic acid, 2-(2-Fluoro-4-biphenyl)propionic acid, 2-(2-Fluoro-4-biphenylyl)propanoic acid, 2-(2-Fluoro-4-biphenylyl)propionic acid, 2-Fluoro-α-methyl-4-biphenylacetic acid, 2-Fluoro-α-methyl-4-diphenylacetic acid, 3-Fluoro-4-phenylhydratropic acid, Adfeed, Ansaid, Antadys, BTS 18322, Cebutid, FP 70, FP-A, Flugalin, Flurbifen, Fluribiprofen, Fluroben, Flurofen, Froben, Racemic flurbiprofen, Stayban, Synalgo, U 27182, Zepolas, dl-2-(2-Fluoro-4-biphenylyl)propionic acid, dl-Flurbiprofen, rac-Flurbiprofen
IUPAC Name 2-(3-fluoro-4-phenylphenyl)propanoic acid
SMILES CC(C(=O)O)c1ccc(c(F)c1)c2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)
Formula Weight 244.09
Percent Purity >95%
Chemical Name or Material Flurbiprofen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.