Learn More
Memmert™ Flush-Fit Set (Frame Covering Gap Between Oven And Wall Opening, W/Air Slots) For Ipp55
Product Code. 16347537 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IPP55 Peltier-cooled Incubator
Unit Size:
Each
Product Code. 16347537
Brand: Memmert™ IPP55B29730
Only null left
Add to basket
Add to basket
Specifications
For Use With (Equipment) IPP55 Peltier-cooled Incubator
Material Stainless Steel
Type Flush-fit Unit