missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone propionate, Thermo Scientific Chemicals

Product Code. 15740919 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15740919 100 mg 100mg
15750919 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15740919 Supplier Thermo Scientific Acros Supplier No. 462101000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80474-14-2
Molecular Formula C25H31F3O5S
Molecular Weight (g/mol) 500.57
InChI Key WMWTYOKRWGGJOA-ZHLGSTKJSA-N
PubChem CID 91820603

Specifications

Melting Point 272°C to 273°C
Quantity 100 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in dioxane
Formula Weight 500.57
Chemical Name or Material Fluticasone propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.