missing translation for 'onlineSavingsMsg'
Learn More

Fluvoxketone, TRC

Product Code. 30010293
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010293 100 mg 100mg
30003977 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010293 Supplier Toronto Research Chemicals Supplier No. TRCF603700100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61718-80-7
Molecular Formula C13 H15 F3 O2
Molecular Weight (g/mol) 260.25
Synonym 5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone, Fluvoxketone, Fluvoxamine Maleate Imp. D (EP)
IUPAC Name 5-methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one
SMILES COCCCCC(=O)c1ccc(cc1)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H15F3O2/c1-18-9-3-2-4-12(17)10-5-7-11(8-6-10)13(14,15)16/h5-8H,2-4,9H2,1H3
Formula Weight 260.1024
Chemical Name or Material Fluvoxketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.