Learn More
Cole-Parmer™ FMI Series RH Valveless Piston Pump Head
Product Code. 15592173 Shop All Masterflex Products
Change view
Click to view available options
Flow Rate:
10 to 100μL
Unit Size:
Each
Product Code. Flow Rate Unit Size
15592173 10 to 100μL Each
1 options
Product Code. 15592173 Supplier Cole-Parmer™ Supplier No. 0711765
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Valveless piston pumps are low maintenance and suffer fewer piston seizures or clogs. With a single moving part, there is less drift over time, offering millions of cycles of reliable, repeatable pumping.
  • Small displacement provides excellent precision
  • Fully adjustable from zero to maximum
Specifications
Description Valveless Piston Pump Head
Flow Rate 10 to 100μL
Maximum Pressure 100 psi