missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-3-(2-naphthyl)-D-alanine, TRC

Product Code. 30179444
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179444 25 g 25g
30179220 1 g 1g
30178939 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179444 Supplier Toronto Research Chemicals Supplier No. TRCF62505025G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138774-94-4
Molecular Formula C28 H23 N O4
Molecular Weight (g/mol) 437.49
Synonym (R)-α-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-2-naphthalenepropanoic Acid
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
SMILES OC(=O)[C@@H](Cc1ccc2ccccc2c1)NC(=O)OCC3c4ccccc4c5ccccc35

Specifications

InChI Formula InChI=1S/C28H23NO4/c30-27(31)26(16-18-13-14-19-7-1-2-8-20(19)15-18)29-28(32)33-17-25-23-11-5-3-9-21(23)22-10-4-6-12-24(22)25/h1-15,25-26H,16-17H2,(H,29,32)(H,30,31)/t26-/m1/s1
Quantity 25 g
Formula Weight 437.163
Shipping Condition Room Temperature
Chemical Name or Material Fmoc-3-(2-naphthyl)-D-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.