missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fmoc-L-Aspartic Acid (OtBu), 10/Pk.

Product Code. 15923264 Shop All Applied Biosystems Products
Change view
Click to view available options
:
433A Peptide Synthesizer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15923264 433A Peptide Synthesizer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15923264 Supplier Applied Biosystems™ Supplier No. 410635

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.

  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each package contains 10 cartridges.
Format Cartridge
For Use With (Equipment) 433 A Peptide Synthesizer
For Use With (Application) Protein Sequencing & Peptide Synthesis
Product Type Peptide Synthesis Reagents
Quantity 10 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.