missing translation for 'onlineSavingsMsg'
Learn More

FMOC-L-Methionine, 98%, Thermo Scientific™

Product Code. 10164630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10164630 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10164630 Supplier Thermo Scientific Acros Supplier No. 265310010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 71989-28-1
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C20H21NO4S
Packaging Glass bottle
Linear Formula FMOC-NHCH(COOH)CH2CH2SCH3
MDL Number MFCD00037134
Specific Rotation -28° to -30° (24°C, 589nm) (c=1, DMF)
Synonym fmoc-met-oh, fmoc-l-methionine, n-fmoc-l-methionine, fmoc-l-met-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, fmoc-met, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine
InChI Key BUBGAUHBELNDEW-UHFFFAOYNA-N
SMILES CSCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid
Specific Rotation Condition −29.00 (24.00°C c=1,DMF)
Molecular Weight (g/mol) 371.45
Quantity 1g
PubChem CID 2724632
Formula Weight 371.45
Percent Purity ≥97.5%
Chemical Name or Material FMOC-L-Methionine, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.