missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fmoc-Val-Cit-PAB-PNP

Product Code. 16463967
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463967 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16463967 Supplier MedChemExpress Supplier No. HY41189
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fmoc-Val-Cit-PAB-PNP is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).

Specifications

Purity 95.87%
CAS 863971-53-3
Content And Storage -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 766.8 g/mol
Product Type Linker
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 40 mg/mL (52.16 mM)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.