Learn More
Foamtec MiraWIPE™ Wipers
Suitable for disinfection of vacuum chambers, medical devices and cleanrooms
POA
Specifications
Product Type MiraWIPE™ Wipes
Material Polyester
View More Specs
Products 2
Product Code Brand Certifications/Compliance Dimensions (L x W) Edges Packaging Quantity Price Quantity & Availability  
Product Code Brand Certifications/Compliance Dimensions (L x W) Edges Packaging Quantity Price Quantity & Availability  
15193504
View Documents Promotion Details
Foamtec
HT5790S
ISO Class 4 9 x 9 in. (22.86 x 22.86 cm) Extra wide sealed 150 Wipers/Bag
POA
Pack of 1200
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15113514
View Documents Promotion Details
Foamtec
HT5790S-25
ISO Class 4 9 x 9 in. (22.86 x 22.86 cm) Extra wide sealed 25 Wipers/Bag
POA
Pack of 1000
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
MiraWIPE™ wipers feature a non-symmetrical micro fiber structure allows them to scrub microbial biofilms and sporicidal residues.
  • Made of continuous filament micro denier, super soft polyester/nylon textile
  • High absorbency and contamination pickup
  • Abrasion resistant, no-run, no-stretch fabric to minimize In-Use Particles from the face of the wiper
  • Available in configurations to meet Class 1-1000 applications
Specifications
Specifications
MiraWIPE™ Wipes
Polyester