missing translation for 'onlineSavingsMsg'
Learn More

Folic acid, 95-102%

Product Code. 15435239
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435239 5 g 5g
15445239 25 g 25g
15455239 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435239 Supplier Thermo Scientific Alfa Aesar Supplier No. J62937.06
 Restricted Item
In Stock
Aggiungi al carrello
Edge
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 59-30-3 | C19H19N7O6 | 441.404 g/mol

Folic acid and its derivatives are essential mediators of one-carbon metabolism within cells.It is especially important in aiding rapid cell division and growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
MDL Number MFCD00079305
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifica

Packaging Glass bottle
Quantity 5 g
Beilstein 100781
Solubility Information Soluble in water (1.6mg/ml),1 M NaOH (50mg/ml),methanol (slightly),diluted acids,and alkaline solutions.
Formula Weight 441.41
Physical Form Powder
Chemical Name or Material Folic acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.