missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid, Fisher BioReagents

Product Code. 11443074
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Packaging:
Amber Glass
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11443074 5 g Amber Glass 5g
11423074 10 g Amber Glass 10g
11433074 25 g Amber Glass 25g
10637165 100 g Amber Glass 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11443074 Supplier Fisher BioReagents Supplier No. BP25195

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Folic Acid is used in cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifications

Melting Point 250°C
Color Yellow to Orange
Density 1.59g/cm³
pH 4.0 to 4.8
Ignition Residue 0.3% max.
Loss on Drying 8.5% max.
Assay Percent Range ≥95.0 %
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 4248
Formula Weight 441.39
Percent Purity 95.0% min.
Physical Form Powder or Solid
Chemical Name or Material Folic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.