missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid, TRC

Product Code. 30010599
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010599 100 g 100g
17919164 1 g 1g
17908504 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010599 Supplier Toronto Research Chemicals Supplier No. TRCF680300100G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19 H19 N7 O6
Molecular Weight (g/mol) 441.4
Synonym Folic Acid, N-[4-[[(2-Amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic acid, Acifolic, Aspol, Cytofol, Dosfolat B activ, Folacid, Folacin, Folbal, Folcidin, Foldine, Folettes, Foliamin, Folicet, Folipac, Folovit, Folsan, Folsaure, Folsav, Folvite, Incafolic, Liver Lactobacillus casei factor, Millafol, NSC 3073, PGA, Pteroyl-L-glutamic acid, Pteroyl-L-monoglutamic acid, Pteroylglutamic acid, Pteroylmonoglutamic acid, Vitamin Bc, Vitamin Be, Vitamin M
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES NC1=NC(=O)c2nc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)cnc2N1

Specifications

InChI Formula InChI=1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1
Quantity 100 g
Formula Weight 441.1397
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Folic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.