missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folpet-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Folpet D4 |
| CAS | 1327204-12-5 |
| Molecular Formula | C9 2H4 Cl3 N O2 S |
| Quantity | 0.01 g |
| Synonym | Folpet D4 |
| InChI Formula | InChI=1 S/C9H4Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2C(=O)N(SC(Cl)(Cl)Cl)C(=O)c2c1[2 H] |
| IUPAC Name | 4,5,6,7-tetradeuterio-2-(trichloromethylsulfanyl)isoindole-1,3-dione |
| Molecular Weight (g/mol) | 300.58 |
| Formula Weight | 298.9279 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction