Learn More
Food Calorimeter Replacement Parts
Product Code. 12443180 Shop All Nickel-Electro Products
Change view
Click to view available options
For Use With (Equipment):
Food Calorimeter
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12443180 Food Calorimeter Each
1 options
Product Code. 12443180 Supplier Nickel Electro Supplier No. 16815
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement parts for Food Calorimeter.
Replacement nickel crucible and igniter coils for Food Calorimeter (Cat. No. S17038).
Specifications
Product Type Nickel Crucible
For Use With (Application) Used for Diluting Alkalis
For Use With (Equipment) Food Calorimeter
Height (English) 2.24 in.
Height (Metric) 5.7 cm