Learn More
Testo™ Food Penetration Probe
Product Code. 10760433
Change view
Click to view available options
Description:
Food Penetration Probe
Unit Size:
Each
Product Code. Description Unit Size
10760433 Food Penetration Probe Each
1 options
Product Code. 10760433 Supplier Testo™ Supplier No. 0603 2492
  • ONLINE EXCLUSIVE PRICE
    247.05 €
    114.00 € / Pack of 1
    Save 133.05 €
    53% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Description Food Penetration Probe
For Use With (Equipment) testo 926, testo 735
Type Thermocouple type T
For Use With (Application) Measuring the core temperature of food
Includes Fixed 1.5 m cable
Length (Metric) 114.3 mm