Learn More
Abeba Food Trax Safety Shoes
Product Code. 32214814
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
Unit Size:
1 pair
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
32214814 42 1 pair
32214808 36 1 pair
32214779 37 1 pair
32214782 38 1 pair
32214777 39 1 pair
32214809 40 1 pair
32214762 41 1 pair
32214781 43 1 pair
32214780 44 1 pair
32214776 45 1 pair
32214798 46 1 pair
32214794 47 1 pair
32214771 48 1 pair
32214778 49 1 pair
32214811 50 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 32214814 Supplier Abeba Supplier No. 501086042
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Safety shoe for use in a variety of applications and industries
  • ESD-Compliant Safety Shoes Designed for Use in ESD-Sensitive Environments
  • S3 Classification Provides Workplace Safety Protection
  • Compliant with DIN EN 61340 ESD Requirements
  • Composite Toe Cap Helps Protect Against Impact and Compression Hazards
  • Antistatic Design Helps Reduce Electrostatic Charge Buildup
  • Black Upper Provides a Professional Appearance
  • Available in Sizes 36 to 50
Specifications
Color Black
Material Microfiber
Size 42
Toe Material Composite
Disposable Reusable
Gender Unisex
Fit Standard Width
Latex Free Yes
Waterproof Water Resistant
EN ISO 20345 S3
Slip Resistance Rating SRC
Product Type Safety Shoes