missing translation for 'onlineSavingsMsg'
Learn More

Manutan Fork extension, W x D 100 x 50mm, extended length 2440mm

Product Code. 12806169
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12806169 Supplier Manutan Supplier No. A338714
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safely lengthens forks for longer loads, held in place by integral retaining bar. Manufactured from heavy duty 8mm thick steel. Conforms to ISO 13284 British standards with test certificate supplied.

  • Effective length (mm): 2440mm
  • Maximum fork (W x D): 100mm x 50mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.