missing translation for 'onlineSavingsMsg'
Learn More

Formal Buffered Saline, Reagecon

Product Code. 11503202 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
HDPE Bottle GL45
HDPE Jerrycan
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11503202 1 L HDPE Bottle GL45 1 L
11593192 5 L HDPE Jerrycan 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11503202 Supplier Reagecon Supplier No. FBS1
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fixative mixture of formaldehyde in isotonic saline for routine histopathology.

  • Fixative for histopathology.

Specifications

Physical Form Liquid
Packaging HDPE Bottle GL45
Quantity 1 L
compliance-icons compliance-icons
Product Identifier
  • FORMAL BUFFERED SALINE
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Formaldehyde, Sodiumdihydrogenphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.