missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde 2,4-Dinitrophenylhydrazone, TRC

Product Code. 30012179
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012179 1 g 1g
30011411 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012179 Supplier Toronto Research Chemicals Supplier No. TRCF6914001G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1081-15-8
Molecular Formula C7 H6 N4 O4
Molecular Weight (g/mol) 210.15
Synonym Formaldehyde, (2,4-dinitrophenyl)hydrazone (6CI,8CI,9CI), Methanal 2,4-dinitrophenylhydrazone, NSC 404128, NSC 48464
IUPAC Name N-(methylideneamino)-2,4-dinitroaniline
SMILES [O-][N+](=O)c1ccc(NN=C)c(c1)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H6N4O4/c1-8-9-6-3-2-5(10(12)13)4-7(6)11(14)15/h2-4,9H,1H2
Formula Weight 210.0389
Percent Purity >95%
Chemical Name or Material Formaldehyde-2,4-dinitrophenylhydrazone (DNPH)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.