missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol

Product Code. 15496105
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496105 4 x 1 L 1 L
11484990 500 mL 500mL
11387638 1 L 1 L
11357638 1 L 1 L
11367638 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15496105 Supplier Thermo Scientific Chemicals Supplier No. 033314.M1
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-00-0 | CH2O | 30.03 g/mol

Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and is used to make components for transmissions, electrical systems, engine blocks, door panels, axles and brake shoes. It serves as a disinfectant. It is also an ingredient in glues and is used as a preservative in medical laboratories, as an embalming fluid and as a sterilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point −15°C
Color Colorless
Density 1.083 g/mL
Boiling Point 97°C
Flash Point 60°C (140°F)
Assay Percent Range 36.5-38.0%
Odor Pungent
Linear Formula HCHO
Refractive Index 1.3765
Quantity 4 x 1 L
UN Number UN1198
Beilstein 1209228
Merck Index 14,4235
Solubility Information Soluble in water.
Formula Weight 30.03
Percent Purity 36.5 to 38.0%
Grade ACS
Physical Form Liquid
Assay Stab. with 10 to 15% methanol
Chemical Name or Material Formaldehyde, For ACS analysis
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde, 37% in aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: formaldehyde, methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.