missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde ACS MP Biomedicals

Product Code. 11460612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460612 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460612 Supplier MP Biomedicals Supplier No. 0219404790
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS grade formaldehyde solution for use as a bactericidal agent and in organic synthesis.

Specifications

CAS 50-00-0
Density 1.03 (air equals 1)
Assay Percent Range 36.5-38%
Molecular Formula CH2O
MDL Number MFCD00003274
Quantity 500 mL
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Molecular Weight (g/mol) 30.03
PubChem CID 712
ChEBI CHEBI:16842
Formula Weight 30.03
Grade ACS
Physical Form Solution
Chemical Name or Material Formaldehyde, ACS, Contains 10-15% methanol.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde, ACS
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H370-Causes damage to organs.
  • H335-May cause respiratory irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301-IF SWALLOWED:.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.