missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol

Product Code. 10722204 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10722204 5 kg Plastic bottle 5kg
10299980 100 g Plastic bottle 100g
10647021 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10722204 Supplier Thermo Scientific Chemicals Supplier No. 410730050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formaldehyde
Name Note 37 wt% Solution
CAS 50-00-0, 7732-18-5, 67-56-1
Melting Point -15.0°C
Color Clear
Density 1.0830g/mL
Boiling Point 97.0°C
Flash Point 50°C
Ignition Residue 0.005% max.
Packaging Plastic bottle
Molecular Formula CH2O
Linear Formula HCHO
Refractive Index 1.3755 to 1.3775
MDL Number MFCD00003274
Quantity 5 kg
Fieser 01,397; 02,200; 04,238; 05,312; 06,264; 07,158; 08,231; 09,224; 10,186; 11,240; 12,232; 14,168; 15,161; 16,171
Merck Index 15, 4263
Specific Gravity 1.083
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and acetone
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Stabilizer 10 to 15% methanol
Molecular Weight (g/mol) 30.03
PubChem CID 712
ChEBI CHEBI:16842
Formula Weight 30.03
Vapor Pressure 2mbar at 20°C
Percent Purity 36.5 to 38.0%
Grade ACS Reagent
Titratable Acid 0.006 meq/g max.
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.