missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde sodium bisulfite, 95%

Product Code. 11423277
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423277 2500 g 2500g
11403277 100 g 100g
11413277 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423277 Supplier Thermo Scientific Alfa Aesar Supplier No. A16194.0E
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 870-72-4 | CH3NaO4S | 134.081 g/mol

Formaldehyde sodium bisulfite, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It also acts as an pharmaceutical intermediate. It can effectively complex metal ion such as Cu, Zn, Pb, co-depositing with nickel ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formaldehyde sodium bisulfite, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It also acts as an pharmaceutical intermediate. It can effectively complex metal ion such as Cu, Zn, Pb, co-depositing with nickel ion.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic, Keep away from acids Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-72-4
Molecular Formula CH3NaO4S
Molecular Weight (g/mol) 134.081
MDL Number MFCD00044664
InChI Key UOULCEYHQNCFFH-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
PubChem CID 2723822
IUPAC Name sodium;hydroxymethanesulfonate
SMILES C(O)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 200°C (dec.)
Odor Odorless
Quantity 2500 g
Sensitivity Hygroscopic
Merck Index 14,4236
Solubility Information It is soluble in water.
Formula Weight 134.09
Percent Purity 95%
Chemical Name or Material Formaldehyde sodium bisulfite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.