missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde sodium bisulfite addition compound, 95%

Product Code. 15159193 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Unit Size:
1kg
5g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15159193 5 kg Plastic Drum 5kg
10193050 5 g Glass Bottle 5g
10397460 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15159193 Supplier Thermo Scientific Acros Supplier No. 150690051
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 870-72-4 | CH3NaO4S | 134.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-72-4
Molecular Formula CH3NaO4S
Molecular Weight (g/mol) 134.081
MDL Number MFCD00044664
InChI Key UOULCEYHQNCFFH-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
PubChem CID 2723822
IUPAC Name sodium;hydroxymethanesulfonate
SMILES C(O)S(=O)(=O)[O-].[Na+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic Drum
Linear Formula HOCH2SO3Na
Quantity 5 kg
Formula Weight 134.08
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material Formaldehyde sodium bisulfite addition compound, 95%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.