missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Sodium Bisulfite, TRC

Product Code. 30011266
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011266 10 g 10g
30008101 100 g 100g
30005115 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011266 Supplier Toronto Research Chemicals Supplier No. TRCF69146010G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-72-4
Molecular Formula CH3NaO4S
Molecular Weight (g/mol) 134.09
Synonym 1-Hydroxymethanesulfonic Acid Sodium Salt (1:1), Hydroxymethanesulfonic Acid Monosodium Salt, Hydroxymethanesulfonic Acid Sodium Salt, Formaldehyde Sulfite Sodium Salt, Formaldehyde-sodium Bisulfite Adduct, Formbis, Hydroxymethanesulfonic Acid Sodium Salt, Monosodium Hydroxymethanesulfonate, Sodium Formaldehyde Bisulfite, Sodium Hydroxymethanesulfonate, Sodium Hydroxymethylsulfonate
SMILES [Na+].OCS(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/CH4O4S.Na/c2-1-6(3,4)5;/h2H,1H2,(H,3,4,5);/q;+1/p-1
Formula Weight 133.965
Percent Purity >95%
Chemical Name or Material Formaldehyde Sodium Bisulfite
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.