missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution (36.5 - 38% in Water) (Stabilized with ~10-15% Methanol), TRC

Product Code. 30408402
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408402 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408402 Supplier Toronto Research Chemicals Supplier No. TRCF69135550ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formaldehyde
CAS 50-00-0
Molecular Formula C H2 O
Quantity 50 mL
Synonym Formaldehyde, Methanal, BFV, F-gen, FM 282, FS 850A, Fannoform, Floguard 1015, Fordor, Formaldehyde-12C, Formalin, Formalin LM, Formalin Taisei, Formalith, Formic aldehyde, Formol, Fyde, Lysoform, Methaldehyde, Methyl aldehyde, Methylene oxide, Morbicid, NSC 298885, Optilyse, Oxomethane, Oxymethylene, Paraform, Superlysoform
InChI Formula InChI=1S/CH2O/c1-2/h1H2
SMILES C=O
IUPAC Name formaldehyde
Molecular Weight (g/mol) 30.03
Formula Weight 30.0106
Concentration or Composition (by Analyte or Components) 36.5-38 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.