missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99.6%, ACS reagent

Product Code. 10627401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627401 100 g Glass Bottle 100g
10219660 500 g EcoSafPak™ Glass Bottle 500g
10617401 1 kg EcoSafPak™ Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). all customers must complete a declaration before delivery.
Product Code. 10627401 Supplier Thermo Scientific Acros Supplier No. 423741000
  • ONLINE EXCLUSIVE PRICE
    22.50 €
    20.60 € / 100g
    Save 1.90 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). all customers must complete a declaration before delivery.

CAS: 75-12-7 | CH3NO | 45.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.0°C to 3.0°C
Density 1.56 (Air = 1.0)
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Assay Percent Range 99.6%
Linear Formula HCONH2
Vapor Pressure Vapor Pressure: 0.08mbar at 20°C
Beilstein 02, 26
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Formula Weight 45.04
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Formamide
Show More Show Less
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.