missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
Description
Formamide, Extra Pure, SLR
Specifications
pH | 4 to 5 |
Melting Point | 2°C |
Boiling Point | 210°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 0.08mbar at 20°C |
Viscosity | 3.75 mPaS at 20°C |
Formula Weight | 45.04g/mol |
Safety and Handling
Product Identifier
- Formamide
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'