missing translation for 'onlineSavingsMsg'
Learn More

Formamidine disulfide dihydrochloride, 97%

Product Code. p-7025027
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439856 5 g 5g
11449856 25 g 25g
11459856 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439856 Supplier Thermo Scientific Alfa Aesar Supplier No. B20331.06
Add to basket
This item is not returnable. View return policy

CAS: 14807-75-1 | C2H8Cl2N4S2 | 223.13 g/mol

Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14807-75-1
Molecular Formula C2H8Cl2N4S2
Molecular Weight (g/mol) 223.13
MDL Number MFCD00012586
InChI Key BFJQSCVWXZOXGK-UHFFFAOYSA-N
Synonym formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride
PubChem CID 24802135
IUPAC Name carbamimidoylsulfanyl carbamimidothioate;dihydrochloride
SMILES [Cl-].[Cl-].[NH3+]C(=N)SSC([NH3+])=N

Specifications

Melting Point ∼175°C (decomposition)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 223.15
Percent Purity 97%
Chemical Name or Material Formamidine disulfide dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.