missing translation for 'onlineSavingsMsg'
Learn More

Formanilide, 98%

Product Code. 11418910
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418910 25 g 25g
11494074 100 g 100g
11404084 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418910 Supplier Thermo Scientific Alfa Aesar Supplier No. A12472.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-70-8 | C7H7NO | 121.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-70-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00003276
InChI Key DYDNPESBYVVLBO-UHFFFAOYSA-N
Synonym formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
PubChem CID 7671
ChEBI CHEBI:42416
IUPAC Name N-phenylformamide
SMILES C1=CC=C(C=C1)NC=O

Specifications

Melting Point 45°C to 49°C
Density 1.14
Boiling Point 271°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 906934
Merck Index 14,4238
Formula Weight 121.14
Percent Purity 98%
Chemical Name or Material Formanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.