missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 98-100%, Certified AR for Analysis, Fisher Chemical™

Product Code. 10559570 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Plastic Coated Glass bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10559570 500 mL Plastic Coated Glass bottle 500mL
10785711 1 L Plastic Coated Glass bottle 1 L
10141570 2.5 L Plastic Coated Glass bottle 2.5 L
10792354 25 L Plastic container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10559570 Supplier Fisher Chemical Supplier No. F/1900/PB08
  • ONLINE EXCLUSIVE PRICE
    48.38 €
    42.20 € / 500mL
    Save 6.18 €
    12% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CH2O2, CAS Number-64-18-6, formylic acid, myrmicyl, acide formique, methanoic acid, hydrogen carboxylic acid, aminic acid, formira, bilorin, collo-bueglatt, formisoton, 2.5L, 101 deg.C, CHEBI:30751, Colorless, 46.02g/mol, BDAGIHXWWSANSR-UHFFFAOYSA-N, formic acid, 8 deg.C, 3297, 46.025 | CAS: 64-18-6 | CH2O2 | 46.025 g/mol

CH2O2, CAS Number-64-18-6, formylic acid, myrmicyl, acide formique, methanoic acid, hydrogen carboxylic acid, aminic acid, formira, bilorin, collo-bueglatt, formisoton, 2.5L, 101 deg.C, CHEBI:30751, Colorless, 46.02g/mol, BDAGIHXWWSANSR-UHFFFAOYSA-N, formic acid, 8 deg.C, 3297, 46.025

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number 3297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

pH 2.1
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Quantity 500 mL
Packaging Plastic Coated Glass bottle
Physical Form Liquid
Vapor Pressure 44mbar at 20°C
Formula Weight 46.02g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Formic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.