missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, 98+%, pure

Product Code. 10796341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
25mL
Product Code. Quantity Packaging unitSize
10796341 2.5 L Plastic bottle 2.5 L
10705031 25 mL Plastic bottle 25mL
10285711 250 mL Plastic bottle 250mL
10071620 1 L Plastic bottle 1 L
11984881 10 L Plastic drum 10 L
5 options
This item is not returnable. View return policy

Product Code. 10796341

Brand: Thermo Scientific Acros 147930025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64-18-6 | CH2O2 | 46.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Assay Percent Range 98+%
Linear Formula HCO2H
Beilstein 02, 8
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Merck Index 15, 4269
Formula Weight 46.02
Percent Purity ≥98.0%
Grade Pure
Density 1.2200g/mL
Quantity 2.5 L
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Liquid
Refractive Index 1.3701 to 1.3721 (20°C, 589nm)
Vapor Pressure 44mbar at 20°C
Chemical Name or Material Formic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.