missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 10797488 Shop All Fisher Chemical Products
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 mL
1 mL
10 Ampules
2 mL
50 mL
Packaging:
Ampule
Ampules
Nalgene™ Poly Bottle
missing translation for 'unitSize'
0.5mL
10 ampoules
1mL
2mL
50mL
This item is not returnable. View return policy

Product Code. 10797488

missing translation for 'mfr': Fisher Chemical A11710X1AMP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable as a mobile phase additive for LC/MS and UHPLC/MS

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
MDL Number 3297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
IUPAC Name formic acid

Specifications

pH 2.1
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Assay Percent Range ≥99.0 %
SMILES OC=O
Molecular Weight (g/mol) 46.03
Formula Weight 46.02g/mol
Percent Purity ≥99.0%
Quantity 10 Ampules
Packaging Ampules
Physical Form Liquid
Vapor Pressure 44mbar at 20°C
Chemical Name or Material Formic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid, OPTIMA LC/MS Grade (99.5%)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Formic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.