missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, ACS, 96+%

Product Code. 11348058
Change view
Click to view available options
Quantity:
500 g
2 kg
4 x 500 g
Unit Size:
2000g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348058 4 x 500 g 2000g
11338058 500 g 500g
11358058 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11348058 Supplier Thermo Scientific Alfa Aesar Supplier No. 036617.57
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64-18-6 | CH2O2 | 46.025 g/mol

Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 7°C to 9°C
Boiling Point 100°C to 101°C
Color Colorless
Assay Percent Range >96+%
Linear Formula HCOOH
UN Number UN1779
Beilstein 1209246
Merck Index 14,4241
Solubility Information Miscible with water. Partially miscible in toluene,xylene and benzene.
Formula Weight 46.03
Percent Purity ≥96%
Grade ACS
Density 1.220 g/mL
Quantity 4 x 500 g
Flash Point 50°C (156°F)
Physical Form Liquid
Refractive Index 1.371
Sensitivity Hygroscopic
Chemical Name or Material Formic acid, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.