missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, 88%, ACS reagent

Product Code. 11904891 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10 L
2.5 L
500mL
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 11904891

Brand: Thermo Scientific Acros 410770100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 64-18-6 | CH2O2 | 46.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 8.0°C
Boiling Point 101.0°C
Assay Percent Range 88%
Linear Formula HCO2H
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine
Formula Weight 46.02
Percent Purity 88%
Grade ACS Reagent
Density 1.2200g/mL
Quantity 10 L
Flash Point 60°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Liquid
Refractive Index 1.3711 to 1.3731
Residue after Evaporation 0.002% max.
Specific Gravity 1.22
Chemical Name or Material Formic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.