missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid : Triethylamine 1:1 Solution, Solstice

Product Code. 15690900 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15690900 100 mL Glass Bottle 100mL
15600910 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15690900 Supplier Honeywell-Fluka Supplier No. 09752100ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for HPLC, 2M:2M concentrate

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-29-5
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00134943
InChI Key PTMFUWGXPRYYMC-UHFFFAOYSA-N
Synonym unii-un4t038p8m, triethylamine formate, teaf, triethylammonium formate, triethylamine, formate, triethylammonium formate mi, triethylammoniumformate, triethylammonium format, hcooh et3n, formic acid triethylamine
PubChem CID 11084032
IUPAC Name N,N-diethylethanamine;formic acid
SMILES CCN(CC)CC.C(=O)O

Specifications

Color Colorless
Linear Formula (CH3CH2)3NH+ HCOO-
UN Number UN1760
Beilstein 4150472
Concentration 2M:2M concentrate
Formula Weight 147.22g/mol
Grade HPLC
Density 1.01g/mL at 20°C
Quantity 100 mL
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Formic Acid: Triethylamine 1:1 Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.