missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid in Water, 0.1%, BAKER ANALYZED LC/MS, J.T. Baker™
Click to view available options
Quantity:
1 L
Packaging:
Glass
Description
High purity solvents/reagents give optimal performance with minimal risk of contaminants that can limit accuracy and productivity, and they maximize the sensitivity and detecting power of instrumentation.
- Application-optimized and function tested to enhance separation power and reproducibility
- For use in more routine LC/MS applications
- Tested for ESI+, UV-VIS absorbance, trace metals, residue after evaporation, assay and overall LC/MS suitability
Specifications
Absorbance | Max. 0.1 at 240nm, Max. 0.01 at 260nm |
CAS | 64-18-6, 7732-18-5 |
Melting Point | 0°C |
Boiling Point | 100°C |
Assay Percent Range | 0.095 - 0.105% |
Molecular Formula | CH2O2 |
Linear Formula | HCOOH |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt |
Solubility Information | Fully miscible in water |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'